Learn More
Cayman Chemical ANTIBODY IMP-1088 500ug
SDP
Supplier:  Cayman Chemical 25366500 UG
Encompass_Preferred
An inhibitor of N-myristoyltransferase (IC50 = 50 = >1,000 nM); blocks production of infectious RV-16 virus in primary hBECs
Catalog No. 50-293-3103