Learn More
Cayman Chemical IMP-1088, Purity: ≥95%, CAS: 2059148-82-0, 5 mg
SDP
Catalog No. 502936495
Encompass_Preferred
An inhibitor of N-myristoyltransferase (IC50 = 50 = >1,000 nM); blocks production of infectious RV-16 virus in primary hBECs
Catalog No. 50-293-6495 Supplier Cayman Chemical Supplier No. 253665 MG
An inhibitor of N-myristoyltransferase (IC50 = 50 = >1,000 nM); blocks production of infectious RV-16 virus in primary hBECs