Learn More
Cayman Chemical IMR-1A, Purity: ≥95%, CAS: 331862-41-0, 100 mg
SDP
Catalog No. 502917505
Encompass_Preferred
An inhibitor of Notch ternary complex formation (IC50 = 0.5 µM) and an active metabolite of IMR-1; binds to the intracellular domain of Notch (Kd = 2.9 µM) and disrupts the protein-protein interactions between Notch and MAM-1 and CSL transcription factor; increases multiciliation in primary choroid plexus cancer cells at 25 µM
Catalog No. 50-291-7505 Supplier Cayman Chemical Supplier No. 37432100 MG
An inhibitor of Notch ternary complex formation (IC50 = 0.5 µM) and an active metabolite of IMR-1; binds to the intracellular domain of Notch (Kd = 2.9 µM) and disrupts the protein-protein interactions between Notch and MAM-1 and CSL transcription factor; increases multiciliation in primary choroid plexus cancer cells at 25 µM