Learn More
Cayman Chemical INNO-8875, Purity: ≥95%, CAS: 871108-05-3, 5 mg
SDP
Catalog No. 502950237
Encompass_Preferred
An adenosine A1 receptor agonist; binds to adenosine A1 receptors (Ki = 0.97 nM) and is >10,000-fold selective for adenosine A1 over A2 receptors; topical application reduces intraocular pressure in a rabbit model of ocular hypertension at 25 µg/eye; decreases atrial refractoriness and heart rate, as well as slows atrioventricuolar nodal conduction, in anesthetized rats from 1-50 µg/kg
Catalog No. 50-295-0237 Supplier Cayman Chemical Supplier No. 333455 MG
An adenosine A1 receptor agonist; binds to adenosine A1 receptors (Ki = 0.97 nM) and is >10,000-fold selective for adenosine A1 over A2 receptors; topical application reduces intraocular pressure in a rabbit model of ocular hypertension at 25 µg/eye; decreases atrial refractoriness and heart rate, as well as slows atrioventricuolar nodal conduction, in anesthetized rats from 1-50 µg/kg