Learn More
Cayman Chemical Iodixanol, Purity: ≥95% (mixture of isomers), CAS: 92339-11-2, 1 mg
SDP
Catalog No. 502913160
Encompass_Preferred
A contrast reagent; induces apoptosis and mitophagy in renal epithelial cells in a rat model of contrast-induced acute kidney injury; has been used as a density gradient medium in the fractionation of plasma lipoproteins
Catalog No. 50-291-3160 Supplier Cayman Chemical Supplier No. 350201 MG
A contrast reagent; induces apoptosis and mitophagy in renal epithelial cells in a rat model of contrast-induced acute kidney injury; has been used as a density gradient medium in the fractionation of plasma lipoproteins