Learn More
Cayman Chemical ANTIBODY IOWH-032 10mg
SDP
Catalog No. 502992522
Encompass_Preferred
A modulator of CFTR; inhibits CFTR in T84 human colon cells (IC50 = 8.51 µM); has rapid inhibitory and slow potentiating effects on human CFTR with apparent Kd values of 6.1 and 0.64 nM, respectively; potentiates currents through ΔF508-CFTR expressed in X. laevis oocytes at 20 nM; inhibits mouse CFTR with an apparent Kd value of 42.9 µM
Catalog No. 50-299-2522 Supplier Cayman Chemical Supplier No. 3027310 MG
A modulator of CFTR; inhibits CFTR in T84 human colon cells (IC50 = 8.51 µM); has rapid inhibitory and slow potentiating effects on human CFTR with apparent Kd values of 6.1 and 0.64 nM, respectively; potentiates currents through ΔF508-CFTR expressed in X. laevis oocytes at 20 nM; inhibits mouse CFTR with an apparent Kd value of 42.9 µM