Learn More
Cayman Chemical IPBT-5CA, Purity: ≥98%, CAS: 306935-41-1, 50 mg
SDP
Catalog No. 502973084
Encompass_Preferred
A selective agonist of GPR109B (HCA3; EC50 = 400 nM) that displays no activity at GPR109A (HCA2)
Catalog No. 50-297-3084 Supplier Cayman Chemical Supplier No. 1522350 MG
A selective agonist of GPR109B (HCA3; EC50 = 400 nM) that displays no activity at GPR109A (HCA2)