Learn More
Cayman Chemical IPG-2 AM, Purity: ≥90%, CAS: 1369302-24-8, 0.5 mg
SDP
Catalog No. 502913525
Encompass_Preferred
A cell-permeable fluorescent potassium indicator; binds to potassium (Kd = 18 mM); ex/em = 525/545 nm, respectively; has been used to kinetically monitor potassium levels in isolated human platelets via flow cytometry
Catalog No. 50-291-3525 Supplier Cayman Chemical Supplier No. 355400.5 MG
A cell-permeable fluorescent potassium indicator; binds to potassium (Kd = 18 mM); ex/em = 525/545 nm, respectively; has been used to kinetically monitor potassium levels in isolated human platelets via flow cytometry