Learn More
Cayman Chemical ANTIBODY IPG-4 AM 500ug
SDP
Catalog No. 502910148
Encompass_Preferred
A cell-permeable fluorescent potassium indicator; has been used to detect intracellular potassium; binds to potassium (Kd = 7 mM); ex/em = 525/545 nm
Catalog No. 50-291-0148 Supplier Cayman Chemical Supplier No. 35541500 UG
A cell-permeable fluorescent potassium indicator; has been used to detect intracellular potassium; binds to potassium (Kd = 7 mM); ex/em = 525/545 nm