Learn More
Cayman Chemical Isoguanine, Purity: ≥95%, CAS: 3373-53-3, 250 mg
SDP
Catalog No. 502986327
Encompass_Preferred
An isomer of guanine; found naturally but also formed through oxidative damage to DNA; mutagenic in mammalian and bacterial cells in vitro
Catalog No. 50-298-6327 Supplier Cayman Chemical Supplier No. 21703250 MG
An isomer of guanine; found naturally but also formed through oxidative damage to DNA; mutagenic in mammalian and bacterial cells in vitro