Learn More
Cayman Chemical Isomaltose, Purity: ≥95%, CAS: 499-40-1, 100 mg
SDP
Catalog No. 502983892
Encompass_Preferred
A disaccharide containing two glucose subunits joined by an α-(1→6) linkage
Catalog No. 50-298-3892 Supplier Cayman Chemical Supplier No. 20797100 MG
A disaccharide containing two glucose subunits joined by an α-(1→6) linkage