Learn More
Cayman Chemical Isosorbide, Purity: ≥95%, CAS: 652-67-5, 50 g
SDP
Catalog No. 502948158
Encompass_Preferred
A dianhydrohexitol; has been used in the synthesis of NO donors with vasodilatory activity and as a monomer or building block in the synthesis of various polymers and functional materials, respectively
Catalog No. 50-294-8158 Supplier Cayman Chemical Supplier No. 3319750 G
A dianhydrohexitol; has been used in the synthesis of NO donors with vasodilatory activity and as a monomer or building block in the synthesis of various polymers and functional materials, respectively