Learn More
Cayman Chemical Itacitinib, Purity: ≥95%, CAS: 1334298-90-6, 1 mg
SDP
Catalog No. 502938271
Encompass_Preferred
A JAK1 inhibitor; >20- and >200-fold selective for JAK1 over JAK2 and JAK3, respectively
Catalog No. 50-293-8271 Supplier Cayman Chemical Supplier No. 275971 MG
A JAK1 inhibitor; >20- and >200-fold selective for JAK1 over JAK2 and JAK3, respectively