Learn More
Cayman Chemical ITI214, Purity: ≥98%, CAS: 1642303-38-5, 5 mg
SDP
Catalog No. 502992470
Encompass_Preferred
A PDE1 inhibitor (Ki = 0.058 nM); selective for PDE1 over PDE2A, -3B, -4A, -5A, -6, -7B, -8A, -9A, -10A, and -11A (Kis = 0.16-18 µM); enhances novel object recognition in rats at 3 and 6 mg/kg; increases prefrontal cortex cAMP and cGMP levels in mice at 50 mg/kg; reverses MK-801-induced decreases in spontaneous alteration in the T-maze continuous alternation task in mice at 0.1, 3, and 10 mg/kg
Catalog No. 50-299-2470 Supplier Cayman Chemical Supplier No. 303765 MG
A PDE1 inhibitor (Ki = 0.058 nM); selective for PDE1 over PDE2A, -3B, -4A, -5A, -6, -7B, -8A, -9A, -10A, and -11A (Kis = 0.16-18 µM); enhances novel object recognition in rats at 3 and 6 mg/kg; increases prefrontal cortex cAMP and cGMP levels in mice at 50 mg/kg; reverses MK-801-induced decreases in spontaneous alteration in the T-maze continuous alternation task in mice at 0.1, 3, and 10 mg/kg