Learn More
Cayman Chemical JFD00244, Purity: ≥95%, CAS: 96969-83-4, 10 mg
SDP
Catalog No. 502971424
Encompass_Preferred
An inhibitor of SIRT2 (IC50= 56.7 µM); induces granulocytic differentiation in the acute promyelocytic leukemia cell line NB4
Catalog No. 50-297-1424 Supplier Cayman Chemical Supplier No. 1464810 MG
An inhibitor of SIRT2 (IC50= 56.7 µM); induces granulocytic differentiation in the acute promyelocytic leukemia cell line NB4