Learn More
Cayman Chemical JG-98, Purity: ≥98%, CAS: 1456551-16-8, 1 mg
SDP
Catalog No. 502918076
Encompass_Preferred
An inhibitor of the Hsp70-BAG-3 protein-protein interaction (IC50 = 1.6 µM); reduces the proliferation of MDA-MB-231 and MCF-7 cells (EC50s = 0.4 and 0.7 µM, respectively) and 22 other cancer cell lines (EC50s = 0.3-4 µM); toxic to MEFs (EC50 = 22 µM); reduces tumor growth in MCF-7 breast cancer and HeLa cervical cancer mouse xenograft models at 3 mg/kg every two days
Catalog No. 50-291-8076 Supplier Cayman Chemical Supplier No. 383371 MG
An inhibitor of the Hsp70-BAG-3 protein-protein interaction (IC50 = 1.6 µM); reduces the proliferation of MDA-MB-231 and MCF-7 cells (EC50s = 0.4 and 0.7 µM, respectively) and 22 other cancer cell lines (EC50s = 0.3-4 µM); toxic to MEFs (EC50 = 22 µM); reduces tumor growth in MCF-7 breast cancer and HeLa cervical cancer mouse xenograft models at 3 mg/kg every two days