Learn More
Cayman Chemical JW 55, Purity: ≥98%, CAS: 664993-53-7, 5 mg
SDP
Catalog No. 502972270
Encompass_Preferred
An inhibitor of the PARP domain of TNKS1 and TNKS2 (IC50s = 1.9 and 0.83 µM, respectively); decreases canonical Wnt signaling in SW480 and HCT15 colon carcinoma cell lines and reduces proliferation of SW480 cells in vitro
Catalog No. 50-297-2270 Supplier Cayman Chemical Supplier No. 162605 MG
An inhibitor of the PARP domain of TNKS1 and TNKS2 (IC50s = 1.9 and 0.83 µM, respectively); decreases canonical Wnt signaling in SW480 and HCT15 colon carcinoma cell lines and reduces proliferation of SW480 cells in vitro