Learn More
Cayman Chemical ANTIBODY JWH 424 25mg
SDP
Catalog No. 502892637
Encompass_Preferred
A soluble adenylyl cyclase inhibitor (IC50 3.3 nM) selective for sAC over AC1 AC2 AC5 AC8 and AC9 at 100 nM a panel of 322 kinases (IC50s >1000 nM for all) and 46 other targets including GPCRs nuclear receptors and ion channels (RC50s >9 UM for all) inhibits capacitation-induced cAMP accumulation in isolated mouse and human sperm at 5 UM reduces sodium bicarbonate-induced increases in mouse and human sperm beat frequency at 5 UM induces temporary infertility in male mice without affecting mating behavior after a single dose of 50 mg/kg
Catalog No. 50-289-2637 Supplier Cayman Chemical Supplier No. 900120325 MG
A soluble adenylyl cyclase inhibitor (IC50 3.3 nM) selective for sAC over AC1 AC2 AC5 AC8 and AC9 at 100 nM a panel of 322 kinases (IC50s >1000 nM for all) and 46 other targets including GPCRs nuclear receptors and ion channels (RC50s >9 UM for all) inhibits capacitation-induced cAMP accumulation in isolated mouse and human sperm at 5 UM reduces sodium bicarbonate-induced increases in mouse and human sperm beat frequency at 5 UM induces temporary infertility in male mice without affecting mating behavior after a single dose of 50 mg/kg