Learn More
Cayman Chemical K22, Purity: ≥98%, CAS: 2141978-86-9, 5 mg
SDP
Catalog No. 502992234
Encompass_Preferred
An antiviral agent; inhibits replication of SARS-CoV, MERS-CoV, HCoV-229E, FCoV, and MHV in Vero cells at 40 µM; inhibits the formation of double membrane vesicles and viral RNA synthesis in HCoV-229E-infected MRC-5 cells; reduces viral titers in cells infected with various Arteriviridae or Torovirinae viruses
Catalog No. 50-299-2234 Supplier Cayman Chemical Supplier No. 315785 MG
An antiviral agent; inhibits replication of SARS-CoV, MERS-CoV, HCoV-229E, FCoV, and MHV in Vero cells at 40 µM; inhibits the formation of double membrane vesicles and viral RNA synthesis in HCoV-229E-infected MRC-5 cells; reduces viral titers in cells infected with various Arteriviridae or Torovirinae viruses