Learn More
Cayman Chemical K6PC-5, Purity: ≥98%, CAS: 756875-51-1, 10 mg
SDP
Catalog No. 502948182
Encompass_Preferred
An SPHK1 activator; activates SPHK1 in F9-12 cell lysates and isolated murine blood; induces calcium oscillations in HaCaT keratinocytes at 25 and 50 µM; inhibits oxygen-glucose deprivation/reoxygenation-induced cell death in SH-SY5Y cells at 10 µM; increases stratum corneum integrity and corneodesmosome density in a mouse model of UVA and UVB radiation-induced photoaging when applied topically at 1%
Catalog No. 50-294-8182 Supplier Cayman Chemical Supplier No. 3407010 MG
An SPHK1 activator; activates SPHK1 in F9-12 cell lysates and isolated murine blood; induces calcium oscillations in HaCaT keratinocytes at 25 and 50 µM; inhibits oxygen-glucose deprivation/reoxygenation-induced cell death in SH-SY5Y cells at 10 µM; increases stratum corneum integrity and corneodesmosome density in a mouse model of UVA and UVB radiation-induced photoaging when applied topically at 1%