Learn More
Cayman Chemical KDM4D-IN-1, Purity: ≥95%, CAS: 2098902-68-0, 5 mg
SDP
Catalog No. 502917599
Encompass_Preferred
An inhibitor of JMJD2D (IC50 = 0.41 µM); selective for JMJD2D over JMJD1B, JMJD2B, and JARID1A (IC50s = >10 µM for all); decreases the colony formation, migration, and invasion of Caki-1 clear cell renal carcinoma cells and 786-O adenocarcinoma cells at 0.5 µM; reduces tumor growth and vessel development in a 786-O mouse xenograft model
Catalog No. 50-291-7599 Supplier Cayman Chemical Supplier No. 374315 MG
An inhibitor of JMJD2D (IC50 = 0.41 µM); selective for JMJD2D over JMJD1B, JMJD2B, and JARID1A (IC50s = >10 µM for all); decreases the colony formation, migration, and invasion of Caki-1 clear cell renal carcinoma cells and 786-O adenocarcinoma cells at 0.5 µM; reduces tumor growth and vessel development in a 786-O mouse xenograft model