Learn More
Cayman Chemical KI-696, Purity: ≥98%, CAS: 1799974-70-1, 10 mg
SDP
Catalog No. 502945661
Encompass_Preferred
A Keap1-Nrf2 protein-protein interaction inhibitor; binds to the Keap1 Kelch domain (Kd = 1.3 nM) and increases the accumulation of Nrf2 in isolated human bronchial epithelial cells in a concentration-dependent manner; increases the expression of NQO1, GCLM, HMOX1, and TXNRD1 in bronchial epithelial cells isolated from patients with COPD (EC50s = 22, 36, 16, and 27 nM, respectively); increases lung expression of Nqo1, Hmox1, and Txnrd1 in rats at 50 µmol/kg
Catalog No. 50-294-5661 Supplier Cayman Chemical Supplier No. 4203910 MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A Keap1-Nrf2 protein-protein interaction inhibitor; binds to the Keap1 Kelch domain (Kd = 1.3 nM) and increases the accumulation of Nrf2 in isolated human bronchial epithelial cells in a concentration-dependent manner; increases the expression of NQO1, GCLM, HMOX1, and TXNRD1 in bronchial epithelial cells isolated from patients with COPD (EC50s = 22, 36, 16, and 27 nM, respectively); increases lung expression of Nqo1, Hmox1, and Txnrd1 in rats at 50 µmol/kg