Learn More
Cayman Chemical Ko 143, Purity: ≥98%, CAS: 461054-93-3, 5 mg
SDP
Catalog No. 502972952
Encompass_Preferred
A potent and selective inhibitor of BCRP, preventing the export of mitoxantrone and topotecan in breast cancer cell lines (EC50s = 23 and 26 nM, respectively); effective in vivo in mice
Catalog No. 50-297-2952 Supplier Cayman Chemical Supplier No. 152155 MG
A potent and selective inhibitor of BCRP, preventing the export of mitoxantrone and topotecan in breast cancer cell lines (EC50s = 23 and 26 nM, respectively); effective in vivo in mice