Learn More
Cayman Chemical KO947, Purity: ≥98%, CAS: 1695533-89-1, 1 mg
SDP
Catalog No. 502938091
Encompass_Preferred
An ERK inhibitor (IC50 = 50-fold selective for ERK over a panel of 450 kinases; reduces RSK phosphorylation in A375 cancer cells containing constitutively active B-RAFV600E (IC50 = 50 = 50-250 nM for all)
Catalog No. 50-293-8091 Supplier Cayman Chemical Supplier No. 292131 MG
An ERK inhibitor (IC50 = 50-fold selective for ERK over a panel of 450 kinases; reduces RSK phosphorylation in A375 cancer cells containing constitutively active B-RAFV600E (IC50 = 50 = 50-250 nM for all)