Learn More
Cayman Chemical KT182, Purity: ≥98%, CAS: 1402612-62-7, 25 mg
SDP
Catalog No. 502970909
Encompass_Preferred
A potent inhibitor of ABHD6 (IC50s = 1.7, 15.1, and 0.24 nM in situ); brain-penetrant
Catalog No. 50-297-0909 Supplier Cayman Chemical Supplier No. 1540425 MG
A potent inhibitor of ABHD6 (IC50s = 1.7, 15.1, and 0.24 nM in situ); brain-penetrant