Learn More
Cayman Chemical KU-0058948, Purity: ≥95%, CAS: 763111-49-5, 10 mg
SDP
Catalog No. 502945431
Encompass_Preferred
A PARP1 and PARP2 inhibitor (IC50s = 3.4 and 1.5 nM, respectively); selective for PARP1 over PARP3, PARP4, and tankyrase (IC50s = 40, 1,200, and >10,000 nM, respectively); selectively inhibits the survival of embryonic stem cells lacking Brca1 or Brca2 over embryonic stem cells expressing wild-type Brca1 or Brca2; decreases the survival and induces apoptosis in P39 and MUTZ-3 AML cells at 1 µM; potentiates MS-275-induced cytotoxicity in patient-derived AML cells at 5 nM; reduces survival in PTEN-deficient HCT116 cells but not those expressing wild-type PTEN or HCT116+/neo
Catalog No. 50-294-5431 Supplier Cayman Chemical Supplier No. 4209810 MG
A PARP1 and PARP2 inhibitor (IC50s = 3.4 and 1.5 nM, respectively); selective for PARP1 over PARP3, PARP4, and tankyrase (IC50s = 40, 1,200, and >10,000 nM, respectively); selectively inhibits the survival of embryonic stem cells lacking Brca1 or Brca2 over embryonic stem cells expressing wild-type Brca1 or Brca2; decreases the survival and induces apoptosis in P39 and MUTZ-3 AML cells at 1 µM; potentiates MS-275-induced cytotoxicity in patient-derived AML cells at 5 nM; reduces survival in PTEN-deficient HCT116 cells but not those expressing wild-type PTEN or HCT116+/neo