Learn More
Cayman Chemical KX2-391, Purity: ≥98%, CAS: 897016-82-9, 100 mg
SDP
Catalog No. 502994640
Encompass_Preferred
A Src kinase inhibitor (IC50 = ~20 nM); selective for Src over PDGFR, EGFR, JAK1, JAK2, Lck, and ZAP-70; induces cell cycle arrest at the G2/M phase and apoptosis in ERα-positive breast cancer cells; inhibits the growth of Huh7, PLC/PRF/5, Hep3B, and HepG2 cells (GI50s = 9, 13, 26, and 60 nM, respectively); decreases spleen weight and the number of splenic leukemia cells in a mouse model of FLT3-ITD-F691L AML
Catalog No. 50-299-4640 Supplier Cayman Chemical Supplier No. 21429100 MG
A Src kinase inhibitor (IC50 = ~20 nM); selective for Src over PDGFR, EGFR, JAK1, JAK2, Lck, and ZAP-70; induces cell cycle arrest at the G2/M phase and apoptosis in ERα-positive breast cancer cells; inhibits the growth of Huh7, PLC/PRF/5, Hep3B, and HepG2 cells (GI50s = 9, 13, 26, and 60 nM, respectively); decreases spleen weight and the number of splenic leukemia cells in a mouse model of FLT3-ITD-F691L AML