Learn More
Cayman Chemical KY 19382, Purity: ≥95%, CAS: 2226664-93-1, 10 mg
SDP
Catalog No. 502915883
Encompass_Preferred
An inhibitor of the CF5-Dvl protein-protein interaction (IC50 = 19 nM); an inhibitor of GSK3β (IC50 = 10 nM); increases expression of the mRNA encoding WISP-1 and Axin2, markers of Wnt pathway activation, in ATDC-5 chondrogenic cells at 100 nM; increases nuclear and cytosolic levels of β-catenin in ATDC-5 cells at 100 nM; increases bone plate growth and tibia length in adolescent mice at 0.1 mg/kg per day
Catalog No. 50-291-5883 Supplier Cayman Chemical Supplier No. 3757910 MG
An inhibitor of the CF5-Dvl protein-protein interaction (IC50 = 19 nM); an inhibitor of GSK3β (IC50 = 10 nM); increases expression of the mRNA encoding WISP-1 and Axin2, markers of Wnt pathway activation, in ATDC-5 chondrogenic cells at 100 nM; increases nuclear and cytosolic levels of β-catenin in ATDC-5 cells at 100 nM; increases bone plate growth and tibia length in adolescent mice at 0.1 mg/kg per day