Learn More
Cayman Chemical KYP-2047, Purity: ≥98%, CAS: 796874-99-2, 25 mg
SDP
Catalog No. 502916906
Encompass_Preferred
A prolyl endopeptidase inhibitor (Ki = 0.02 nM for the porcine enzyme); inhibits hydrogen peroxide-induced aggregation of α-synuclein in SH-SY5Y cells expressing wild-type, α-synucleinA30P, or α-synucleinA53T in in vitro models of Parkinson’s disease at 1 µM; decreases palmitic acid-induced lipid peroxidation and ROS production in HepG2 cells at 10 µM; decreases ileal and pulmonary edema and pulmonary fibrosis in a mouse model of acute lung injury induced by intestinal ischemia-reperfusion at 2.5 mg/kg; reduces tumor volume and intratumoral levels of Vegf and Cd34 in a U87 glioblastoma mouse xenograft model at 2.5 mg/kg
Catalog No. 50-291-6906 Supplier Cayman Chemical Supplier No. 3918025 MG
A prolyl endopeptidase inhibitor (Ki = 0.02 nM for the porcine enzyme); inhibits hydrogen peroxide-induced aggregation of α-synuclein in SH-SY5Y cells expressing wild-type, α-synucleinA30P, or α-synucleinA53T in in vitro models of Parkinson’s disease at 1 µM; decreases palmitic acid-induced lipid peroxidation and ROS production in HepG2 cells at 10 µM; decreases ileal and pulmonary edema and pulmonary fibrosis in a mouse model of acute lung injury induced by intestinal ischemia-reperfusion at 2.5 mg/kg; reduces tumor volume and intratumoral levels of Vegf and Cd34 in a U87 glioblastoma mouse xenograft model at 2.5 mg/kg