Learn More
Cayman Chemical L-778,123, Purity: ≥95%, CAS: 183499-57-2, 10 mg
SDP
Catalog No. 502949000
Encompass_Preferred
A dual inhibitor of FTase and GGTase I (IC50s = 2 and 98 nM, respectively); inhibits prenylation of FTase and GGTase I substrates HDJ2 and Rap1a in PSN-1 pancreatic tumor cells (EC50s = 92 and 6,760 nM, respectively); inhibits prenylation of the oncogenic protein Ki-Ras in PSN-1 cells in a concentration-dependent manner; reduces HDJ2 and Rap1a prenylation ex vivo in dog PBMCs when administered at a dose of 35-50 mg/kg per day; has no effect on Ki-Ras prenylation in patient-derived PBMCs; inhibits lectin-induced expression of the T cell activation markers CD71 and CD25 on human PMBCs (IC50s = 6.48 and 84.1 μM, respectively); inhibits IL-2-induced proliferation of CTLL2 cells (IC50 = 0.81 μM)
Catalog No. 50-294-9000 Supplier Cayman Chemical Supplier No. 3250410 MG
A dual inhibitor of FTase and GGTase I (IC50s = 2 and 98 nM, respectively); inhibits prenylation of FTase and GGTase I substrates HDJ2 and Rap1a in PSN-1 pancreatic tumor cells (EC50s = 92 and 6,760 nM, respectively); inhibits prenylation of the oncogenic protein Ki-Ras in PSN-1 cells in a concentration-dependent manner; reduces HDJ2 and Rap1a prenylation ex vivo in dog PBMCs when administered at a dose of 35-50 mg/kg per day; has no effect on Ki-Ras prenylation in patient-derived PBMCs; inhibits lectin-induced expression of the T cell activation markers CD71 and CD25 on human PMBCs (IC50s = 6.48 and 84.1 μM, respectively); inhibits IL-2-induced proliferation of CTLL2 cells (IC50 = 0.81 μM)