Learn More
Cayman Chemical L-Abrine, Purity: ≥98%, CAS: 526-31-8, 100 mg
SDP
Catalog No. 502910352
Encompass_Preferred
A methylated derivative of L-tryptophan; promotes growth in rats fed a tryptophan-deficient diet; unlike tryptophan, it does not affect sleep or wakefulness in rats; serum levels are decreased in patients after an acute ischemic stroke; increased urine levels have been used as a biomarker for abrin poisoning
Catalog No. 50-291-0352 Supplier Cayman Chemical Supplier No. 34914100 MG
A methylated derivative of L-tryptophan; promotes growth in rats fed a tryptophan-deficient diet; unlike tryptophan, it does not affect sleep or wakefulness in rats; serum levels are decreased in patients after an acute ischemic stroke; increased urine levels have been used as a biomarker for abrin poisoning