Learn More
Cayman Chemical L-Proline, Purity: ≥95%, CAS: 147-85-3, 1 kg
SDP
Catalog No. 502989620
Encompass_Preferred
A nonessential amino acid; a substrate for PAT1; an inhibitor of AChE (Ki = 86 µM); accumulates in plants under environmental stress and is important for tolerance to environmental stress
Catalog No. 50-298-9620 Supplier Cayman Chemical Supplier No. 307721 KG
A nonessential amino acid; a substrate for PAT1; an inhibitor of AChE (Ki = 86 µM); accumulates in plants under environmental stress and is important for tolerance to environmental stress