Learn More
Cayman Chemical L-Proline, Purity: ≥95%, CAS: 147-85-3, 250 g
SDP
Catalog No. 502988193
Encompass_Preferred
A nonessential amino acid; a substrate for PAT1; an inhibitor of AChE (Ki = 86 µM); accumulates in plants under environmental stress and is important for tolerance to environmental stress
Catalog No. 50-298-8193 Supplier Cayman Chemical Supplier No. 30772250 G
A nonessential amino acid; a substrate for PAT1; an inhibitor of AChE (Ki = 86 µM); accumulates in plants under environmental stress and is important for tolerance to environmental stress