Learn More
Aat Bioquest ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction*
SDP
Catalog No. 501953008
Encompass_Preferred
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications. These dyes have large Stokes Shifts, and can be well excited by the laser lines of flow cytometers (e.g., 405 nm, 488 nm and 633 nm).
Catalog No. 50-195-3008 Supplier Aat Bioquest Supplier No. 1120
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications. These dyes have large Stokes Shifts, and can be well excited by the laser lines of flow cytometers (e.g., 405 nm, 488 nm and 633 nm).