Learn More
Fluidigm Corp ANTIBODY LABELING KIT 148ND
SDP
Catalog No. NC0260968
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 148Nd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0260968 Supplier Fluidigm Corp Supplier No. 201148A
This Maxpar kit lets you conjugate IgG antibodies with the 148Nd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.