Learn More
Aat Bioquest Buccutite™ Rapid APC Antibody Labeling Kit *Microscale Optimized for Labeling 100 ug Antibody Per Reaction*
SDP
Catalog No. 501953033
Encompass_Preferred
APC is an red fluorescent protein which has an excitation wavelength of 651 nm and an emission wavelength of 662 nm.
Catalog No. 50-195-3033 Supplier Aat Bioquest Supplier No. 1311
APC is an red fluorescent protein which has an excitation wavelength of 651 nm and an emission wavelength of 662 nm.