Learn More
Invitrogen™ Antibody Labeling Kits for 100 μg
Description
Alexa Fluor antibody labeling kits are designed for efficiency and reliability, delivering ready-to-use labeled proteins in just 30 minutes, with minimal hands-on time. The kits have been redeveloped with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of pure conjugated antibody.
Alexa Fluor antibody labeling kit features include:
- Fast and convenient—labeled proteins ready for use in 30 minutes, with less than 15 minutes of hands-on time
- High efficiency—label 20 to 100 μg of protein with bright, photostable Alexa Fluor dyes, with three reactions per kit
- Exceptional recovery—pre-packed spin columns for rapid removal of unbound dye and high conjugate yield
- Comprehensive guidance—includes detailed instructions and a convenient calculator for determining the degree of labeling (DOL)
- Versatile applications—suitable for labeling a variety of purified proteins, including antibodies, growth factors, cytokines, nanobodies, and enzymes
Superior performance and a faster workflow
Alexa Fluor antibody labeling kits deliver superior results with lower background fluorescence, reduced nonspecific binding, and streamlined workflows. Each kit is equipped with materials for three antibody conjugation reactions with one of ten different fluorophores or biotin. Bright, photostable Alexa Fluor dyes react efficiently with primary amines of proteins to form stable dye–protein conjugates. Removal of free dye with the included spin columns enables accurate determination of dye‐to‐protein ratios.
The Alexa Fluor antibody labeling protocol is simple and rapid, ensuring minimal hands-on time and reliable results. The kits include all necessary reagents for antibody labeling, including amine-reactive dye, buffer, and purification columns. The reactive dyes contained in these kits readily react with lysine residues in antibodies or proteins to yield a covalently attached fluorescence-based sensor. Spin columns included in the kits are used for purifying the labeled antibody from excess dye with yields of 70–95%. The kits contain sufficient reagents for three labeling reactions of 20 to 100 μg of antibody. The wide variety of colors are compatible with various detection systems, including fluorescence microscopy, flow cytometry, and high content imaging platforms.
Specifications
Specifications
| For Use With (Application) | Antibody and Protein Labeling |
| Product Type | Labeling Kit |
| Quantity | 3 x 100 μg |
| Detection Method | Fluorescence |
| Excitation/Emission | 590/616 nm |
| Product Line | Alexa Fluor |
| Chemical Reactivity | Amine |
| Solubility | Aqueous Buffer |
| Label or Dye | Alexa Fluor 594 |
| Labeling Target | Antibodies |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.