Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Antibody Labeling Kits for 100 μg

Catalog No. A88071
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Alexa Fluor 350
Alexa Fluor 488
Alexa Fluor 532
Alexa Fluor 546
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 680
Alexa Fluor 790
Biotin
None
Quantity:
3 x 0.5 to 1 mg
3 x 100 μg
3 x 20 to 100 μg
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Quantity
A88071 None 3 x 20 to 100 μg
A88072 None 3 x 0.5 to 1 mg
B88060 Biotin 3 x 100 μg
A88061 Alexa Fluor 350 3 x 100 μg
A88062 Alexa Fluor 488 3 x 100 μg
A88063 Alexa Fluor 532 3 x 100 μg
A88064 Alexa Fluor 546 3 x 100 μg
A88065 Alexa Fluor 555 3 x 100 μg
A88066 Alexa Fluor 568 3 x 100 μg
A88067 Alexa Fluor 594 3 x 100 μg
A88068 Alexa Fluor 647 3 x 100 μg
A88069 Alexa Fluor 680 3 x 100 μg
A88070 Alexa Fluor 790 3 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. A88071 Supplier Invitrogen™ Supplier No. A88071
Only null left
Add to Cart
Add to Cart

Alexa Fluor™ Antibody Labeling Kits are designed for efficiency and reliability, delivering ready-to-use labeled proteins in just 30 minutes, with minimal hands-on time. The kits have been redeveloped with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of pure conjugated antibody.

Alexa Fluor antibody labeling kits are designed for efficiency and reliability, delivering ready-to-use labeled proteins in just 30 minutes, with minimal hands-on time. The kits have been redeveloped with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of pure conjugated antibody.

Alexa Fluor antibody labeling kit features include:

  • Fast and convenient—labeled proteins ready for use in 30 minutes, with less than 15 minutes of hands-on time
  • High efficiency—label 20 to 100 μg of protein with bright, photostable Alexa Fluor dyes, with three reactions per kit
  • Exceptional recovery—pre-packed spin columns for rapid removal of unbound dye and high conjugate yield
  • Comprehensive guidance—includes detailed instructions and a convenient calculator for determining the degree of labeling (DOL)
  • Versatile applications—suitable for labeling a variety of purified proteins, including antibodies, growth factors, cytokines, nanobodies, and enzymes

Superior performance and a faster workflow

Alexa Fluor antibody labeling kits deliver superior results with lower background fluorescence, reduced nonspecific binding, and streamlined workflows. Each kit is equipped with materials for three antibody conjugation reactions with one of ten different fluorophores or biotin. Bright, photostable Alexa Fluor dyes react efficiently with primary amines of proteins to form stable dye–protein conjugates. Removal of free dye with the included spin columns enables accurate determination of dye‐to‐protein ratios.

The Alexa Fluor antibody labeling protocol is simple and rapid, ensuring minimal hands-on time and reliable results. The kits include all necessary reagents for antibody labeling, including amine-reactive dye, buffer, and purification columns. The reactive dyes contained in these kits readily react with lysine residues in antibodies or proteins to yield a covalently attached fluorescence-based sensor. Spin columns included in the kits are used for purifying the labeled antibody from excess dye with yields of 70–95%. The kits contain sufficient reagents for three labeling reactions of 20 to 100 μg of antibody. The wide variety of colors are compatible with various detection systems, including fluorescence microscopy, flow cytometry, and high content imaging platforms.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody and Protein Labeling
Product Type Labeling Kit
Quantity 3 x 20 to 100 μg
Detection Method Fluorescence
Product Line Alexa Fluor
Chemical Reactivity Amine
Solubility Aqueous Buffer
Label or Dye None
Labeling Target Antibodies

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.