Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF555 1/EA
Catalog No. 5000499691
Encompass_Preferred
Mix-n-Stain Maxi Antibody Labeling Kits allow you to label 1 mg of antibody with one of our superior CF dyes in 30 minutes without a purification step Orange fluorescent CF555 dye has Ex/Em at 555/565 nm One labeling of 1 mg IgG antibody per kit
This item is not returnable. View return policy
Catalog No. 50-004-99691 Supplier Biotium Supplier No. 92406
Mix-n-Stain Maxi Antibody Labeling Kits allow you to label 1 mg of antibody with one of our superior CF dyes in 30 minutes without a purification step Orange fluorescent CF555 dye has Ex/Em at 555/565 nm One labeling of 1 mg IgG antibody per kit