Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF568 1/EA
Supplier:  Biotium 92587
Encompass_Preferred
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Red fluorescent CF568 has Ex/Em at 562/583 nm One labeling of 5-20 ug Nanobody Thiol per kit
Catalog No. 50-005-01459