Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF680 1/EA
Catalog No. 5000501550
Encompass_Preferred
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Far-red/near-IR CF680 dye has Ex/Em at 681/698 nm One labeling of 20-50 ug Nanobody Thiol per kit
Catalog No. 50-005-01550 Supplier Biotium Supplier No. 92596
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Far-red/near-IR CF680 dye has Ex/Em at 681/698 nm One labeling of 20-50 ug Nanobody Thiol per kit