Learn More
Biotium Antibody Labeling Kits Antibody and Protein Labeling Kits Mix n Stain Antibody Labeling Kits CF740 1/EA
Catalog No. 5000501452
Encompass_Preferred
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Near-IR CF740 dye has Ex/Em at 742/767 nm One labeling of 20-50 ug Nanobody Thiol per kit
Catalog No. 50-005-01452 Supplier Biotium Supplier No. 92598
Mix-n-Stain CF Dye Nanobody Thiol Labeling Kits are optimized for labeling nanobodies with bright and photostable CF dyes in 2 hours with no purification Kits tolerate common stabilizers Near-IR CF740 dye has Ex/Em at 742/767 nm One labeling of 20-50 ug Nanobody Thiol per kit