Learn More
Cayman Chemical LATS-IN-1, Purity: ≥98%, CAS: 1424635-83-5, 1 mg
SDP
Catalog No. 502909497
Encompass_Preferred
An inhibitor of LATS1 and LATS2 (IC50s = 2 nM for both); inhibits phosphorylation of YAP1 in serum-starved HEK293A cells (EC50 = 510 nM); increases the proliferation of Müller glial cells and isolated mouse cardiomyocytes at 10 µM
Catalog No. 50-290-9497 Supplier Cayman Chemical Supplier No. 366231 MG
An inhibitor of LATS1 and LATS2 (IC50s = 2 nM for both); inhibits phosphorylation of YAP1 in serum-starved HEK293A cells (EC50 = 510 nM); increases the proliferation of Müller glial cells and isolated mouse cardiomyocytes at 10 µM