Learn More
Cayman Chemical ANTIBODY LCL-521 10mg
SDP
Catalog No. 502988363
Encompass_Preferred
An inhibitor of acid ceramidase; inhibits acid ceramidase activity in MCF-7 cell at 1 µM; decreases sphingosine levels in MCF-7 cells at 1 and 10 µM; inhibits proliferation of MCF-7 cells (IC50 = 7.46 µM); induces cell cycle arrest at the G1 phase at 2.5 µM
Catalog No. 50-298-8363 Supplier Cayman Chemical Supplier No. 2973810 MG
An inhibitor of acid ceramidase; inhibits acid ceramidase activity in MCF-7 cell at 1 µM; decreases sphingosine levels in MCF-7 cells at 1 and 10 µM; inhibits proliferation of MCF-7 cells (IC50 = 7.46 µM); induces cell cycle arrest at the G1 phase at 2.5 µM