Learn More
Cayman Chemical LCL-521, Purity: ≥95%, CAS: 1226851-11-1, 1 mg
SDP
Catalog No. 502990530
Encompass_Preferred
An inhibitor of acid ceramidase; inhibits acid ceramidase activity in MCF-7 cell at 1 µM; decreases sphingosine levels in MCF-7 cells at 1 and 10 µM; inhibits proliferation of MCF-7 cells (IC50 = 7.46 µM); induces cell cycle arrest at the G1 phase at 2.5 µM
Catalog No. 50-299-0530 Supplier Cayman Chemical Supplier No. 297381 MG
An inhibitor of acid ceramidase; inhibits acid ceramidase activity in MCF-7 cell at 1 µM; decreases sphingosine levels in MCF-7 cells at 1 and 10 µM; inhibits proliferation of MCF-7 cells (IC50 = 7.46 µM); induces cell cycle arrest at the G1 phase at 2.5 µM