Learn More
Cayman Chemical LCS3, Purity: ≥98%, CAS: 109844-92-0, 1 mg
SDP
Catalog No. 502910077
Encompass_Preferred
An inhibitor of GR and TrxR (IC50s = 3.3 and 3.8 µM, respectively); decreases the GSH/GSSG ratio in NCI H1650 and H23 lung adenocarcinoma cells; induces apoptosis in NCI H1650 and H23 cells but not non-transformed HPL1D lung epithelial cells at 3 µM; active against M. tuberculosis (MIC = 0.098 µM)
Catalog No. 50-291-0077 Supplier Cayman Chemical Supplier No. 366361 MG
An inhibitor of GR and TrxR (IC50s = 3.3 and 3.8 µM, respectively); decreases the GSH/GSSG ratio in NCI H1650 and H23 lung adenocarcinoma cells; induces apoptosis in NCI H1650 and H23 cells but not non-transformed HPL1D lung epithelial cells at 3 µM; active against M. tuberculosis (MIC = 0.098 µM)