Learn More
Cayman Chemical ANTIBODY LGK974 500ug
SDP
Catalog No. 502955385
Encompass_Preferred
A potent and specific small molecule inhibitor of PORCN (IC50 = 0.4 nM); greatly reduces Wnt secretion without decreasing its synthesis and without increasing cytotoxicity; orally bioavailable in mice
Catalog No. 50-295-5385 Supplier Cayman Chemical Supplier No. 14072500 UG
A potent and specific small molecule inhibitor of PORCN (IC50 = 0.4 nM); greatly reduces Wnt secretion without decreasing its synthesis and without increasing cytotoxicity; orally bioavailable in mice