Learn More
Cayman Chemical Linderane, Purity: ≥98%, CAS: 13476-25-0, 25 mg
SDP
Catalog No. 502937846
Encompass_Preferred
A sesquiterpene; inhibits CYP2C9 (Ki = 1.26 μM); reduces Pck1 and G6pc gene expression, cAMP concentration, and CREB phosphorylation, increases PDE3 activity, and inhibits gluconeogenesis in rat primary hepatocytes; decreases serum and hepatic triglyceride levels, as well as random-fed and fasting blood glucose levels in ob/ob mice
Catalog No. 50-293-7846 Supplier Cayman Chemical Supplier No. 2747925 MG
A sesquiterpene; inhibits CYP2C9 (Ki = 1.26 μM); reduces Pck1 and G6pc gene expression, cAMP concentration, and CREB phosphorylation, increases PDE3 activity, and inhibits gluconeogenesis in rat primary hepatocytes; decreases serum and hepatic triglyceride levels, as well as random-fed and fasting blood glucose levels in ob/ob mice