Learn More
Cayman Chemical Linuron, Purity: ≥95%, CAS: 330-55-2, 100 mg
SDP
Catalog No. 502912873
Encompass_Preferred
A phenylurea herbicide; inhibits the androgenic effect of dihydrotestosterone in MDA-kb2 breast cancer cells (IC20 = 1.74 µM in a reporter assay); inhibits testosterone-induced growth of the seminal vesicles, ventral prostate, and levator ani and bulbocavernosus muscles in castrated rats at 100 mg/kg; post-emergent application to carrots controls the growth of the weeds redroot pigweed and lambsquarters
Catalog No. 50-291-2873 Supplier Cayman Chemical Supplier No. 36205100 MG
A phenylurea herbicide; inhibits the androgenic effect of dihydrotestosterone in MDA-kb2 breast cancer cells (IC20 = 1.74 µM in a reporter assay); inhibits testosterone-induced growth of the seminal vesicles, ventral prostate, and levator ani and bulbocavernosus muscles in castrated rats at 100 mg/kg; post-emergent application to carrots controls the growth of the weeds redroot pigweed and lambsquarters