Learn More
Cayman Chemical Lipid 10, Purity: ≥95%, CAS: 2430034-02-7, 25 mg
SDP
Catalog No. 502916370
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.2-6.5); LNPs containing lipid 10 and encapsulating CKAP5 siRNA reduce its gene expression and protein levels in vitro and selectively reduce cell viability in a variety of genetically unstable cancer cell lines; LNPs containing lipid 10 and encapsulating CKAP5 siRNA reduce tumor growth and increase survival in an NCI/ADR-RES human ovarian cancer mouse xenograft model
Catalog No. 50-291-6370 Supplier Cayman Chemical Supplier No. 3870525 MG
An ionizable cationic lipid (pKa = 6.2-6.5); LNPs containing lipid 10 and encapsulating CKAP5 siRNA reduce its gene expression and protein levels in vitro and selectively reduce cell viability in a variety of genetically unstable cancer cell lines; LNPs containing lipid 10 and encapsulating CKAP5 siRNA reduce tumor growth and increase survival in an NCI/ADR-RES human ovarian cancer mouse xenograft model